Anti-MYL7 Rabbit Polyclonal Antibody
Catalog # 77743-756
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MYL7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q01449
- Antigen synonyms:zgc:92755|atrial isoform|cmlc2|MGC92755|MLC-2a|MLC2a|MLCK|cardiac|MLRA_HUMAN|MYL2A|MYL7|Mylc2a|Myosin light chain 2a|Myosin light chain 7 regulatory|Myosin light polypeptide 7 regulatory|Myosin regulatory light chain 2|Myosin regulatory light chain 2 atrial isoform|Myosin regulatory light chain 7
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Myosin regulatory light chain 7
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MYL7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat