Anti-GPT2 Rabbit Polyclonal Antibody
Catalog # 77738-939
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:GPT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8TD30
- Antigen synonyms:AAT2|Alanine aminotransferase 2|ALAT2_HUMAN|ALT2|Glutamate pyruvate transaminase 2|Glutamic alanine transaminase 2|Glutamic pyruvate transaminase (alanine aminotransferase) 2|Glutamic pyruvic transaminase 2|Glutamic--alanine transaminase 2|Glutamic--pyruvic transaminase 2|GPT 2|gpt2
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:AAT2
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: GPT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat