Anti-HUS1 Rabbit Polyclonal Antibody
77740-538
New Product
- Antigen symbol:HUS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O60921
- Antigen synonyms:Checkpoint protein hHUS1 (S. pombe) checkpoint homolog|HUS1 checkpoint homolog (S. pombe)|HUS1 Checkpoint Protein|HUS1 protein|HUS1+ - like protein|HUS1_HUMAN|Hydroxyurea-sensitive 1, S. pombe, homolog of
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Hydroxyurea-sensitive 1, S. pombe, homolog of
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: HUS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat