Anti-FUCA1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: FUCA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-645
- 77737-709
- Antigen symbol:
- FUCA1
- FUCA1
- Antigen synonyms:
- Alpha L fucosidase I|Alpha L fucoside fucohydrolase|Alpha-L-fucosidase 1|Alpha-L-fucosidase I|Alpha-L-fucoside fucohydrolase 1|FUCA|FUCO_HUMAN|fucosidase|alpha-L|tissue|alpha-L- 1|Tissue alpha L fucosidase|Tissue alpha-L-fucosidase
- alpha-L- 1|fucosidase|Alpha-L-fucosidase 1|FUCO_HUMAN|tissue|FUCA|Alpha L fucoside fucohydrolase|Alpha-L-fucosidase I|Alpha-L-fucoside fucohydrolase 1|Tissue alpha-L-fucosidase|Tissue alpha L fucosidase|Alpha L fucosidase I|alpha-L
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P04066
- P04066
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Tissue alpha-L-fucosidase protein (32-158AA)
- Recombinant Human Tissue alpha-L-fucosidase protein (32-158AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together