Anti-GOSR1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: GOSR1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-264
- 77738-681
- Antigen symbol:
- GOSR1
- GOSR1
- Antigen synonyms:
- GOSR1|GS28Golgi SNAP receptor complex member 1|28 kDa Golgi SNARE protein|28 kDa cis-Golgi SNARE p28|GOS-28
- GOSR1|GS28Golgi SNAP receptor complex member 1|28 kDa Golgi SNARE protein|28 kDa cis-Golgi SNARE p28|GOS-28
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95249
- O95249
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Golgi SNAP receptor complex member 1 protein (1-115AA)
- Recombinant Human Golgi SNAP receptor complex member 1 protein (1-115AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together