Anti-HIST1H2BB Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HIST1H2BB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-045
- 77739-815
- Antigen symbol:
- HIST1H2BB
- Antigen synonyms:
- HIST1H2BB|H2BFFHistone H2B type 1-B|Histone H2B.1|Histone H2B.f|H2B/f
- HIST1H2BB|H2BFFHistone H2B type 1-B|Histone H2B.1|Histone H2B.f|H2B/f
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P33778
- P33778
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Histone H2B type 1-B protein (2-126AA)
- Recombinant Human Histone H2B type 1-B protein (2-126AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together