Anti-RIPOR2 Rabbit Recombinant polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: RIPOR2
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77736-727
- 77855-177
- Antigen symbol:
- RIPOR2
- RIPOR2
- Antigen synonyms:
- Ab2 162| C6orf32| DIFF48| FA65B_HUMAN| Fam65b| KIAA0386| PL48| Protein FAM65B
- Ab2 162|C6orf32|DIFF48|FA65B_HUMAN|Fam65b|KIAA0386|PL48|Protein FAM65B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y4F9
- Q9Y4F9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Rho family-interacting cell polarization regulator 2 protein (1-250AA)
- Recombinant Human Rho family-interacting cell polarization regulator 2 protein (1-250AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together