Anti-MYO1B Rabbit Polyclonal Antibody
Catalog # 77743-774
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MYO1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:100 µl
- Form:Liquid
- Gene ID:O43795
- Antigen synonyms:MYO1B_HUMAN|Myosin I alpha|OTTHUMP00000165861|MYH-1c|MMI-alpha|myosin IB|myr1|Unconventional myosin Ib|MYH 1c|MMI alpha|MMIa|MYO1B variant protein|Myosin-Ib
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Unconventional myosin Ib
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MYO1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat