Anti-KCNE1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: KCNE1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-464
- 77858-418
- Antigen symbol:
- KCNE1
- KCNE1
- Antigen synonyms:
- Delayed rectifier potassium channel subunit IsK|Human cardiac delayed rectifier potassium channel protein|IKs producing slow voltage gated potassium channel subunit beta Mink|IKs producing slow voltage-gated potassium channel subunit beta Mink|ISK|JLNS 2|JLNS|JLNS2|KCNE 1|KCNE1_HUMAN|LQT 5|LQT5|MGC33114|Minimal potassium channel|MinK|Potassium voltage gated channel Isk related family member 1|Potassium voltage gated channel Isk related subfamily member 1|Potassium voltage gated channel subfamily E member 1|Potassium voltage-gated channel subfamily E member 1
- Delayed rectifier potassium channel subunit IsK|Human cardiac delayed rectifier potassium channel protein|IKs producing slow voltage gated potassium channel subunit beta Mink|IKs producing slow voltage-gated potassium channel subunit beta Mink|ISK|JLNS 2|JLNS|JLNS2|KCNE 1|KCNE1_HUMAN|LQT 5|LQT5|MGC33114|Minimal potassium channel|MinK|Potassium voltage gated channel Isk related family member 1|Potassium voltage gated channel Isk related subfamily member 1|Potassium voltage gated channel subfamily E member 1|Potassium voltage-gated channel subfamily E member 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P15382
- P15382
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Potassium voltage-gated channel subfamily E member 1 protein (85-129AA)
- Recombinant Human Potassium voltage-gated channel subfamily E member 1 protein (85-129AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together