Anti-SLC9A8 Rabbit Polyclonal Antibody
Catalog # 77736-788
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:SLC9A8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9Y2E8
- Antigen synonyms:Na(+)/H(+) exchanger 8|NHE 8|NHE-8|NHE8|SL9A8_HUMAN|Sodium/ hydrogen exchanger 8|Sodium/hydrogen exchanger 8|solute carrier family 9 (sodium/hydrogen exchanger)|member 8|Solute carrier family 9 member 8
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Na(+)/H(+) exchanger 8
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, IHC: 1:200 to 1:1000, ELISA:1:40000
Type:
Antigen: SLC9A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat