Order Entry
Canada
ContactUsLinkComponent
Anti-NME6 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-NME6 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
Anti-NME6 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77744-564
  • 77744-565
  • Antigen symbol:
  • NME6
  • NME6
  • Antigen synonyms:
  • Inhibitor of p53 induced apoptosis alpha|Inhibitor of p53-induced apoptosis-alpha|IPIA ALPHA|IPIA-alpha|NDK 6|NDK6_HUMAN|NDP kinase 6|NM23 H6|nm23-H6|NM23H6|Nme6|Nucleoside diphosphate kinase 6|OTTHUMP00000164810|OTTHUMP00000210736|OTTHUMP00000210737|OTTHUMP00000210738|OTTHUMP00000210741|OTTHUMP00000210742|OTTHUMP00000210744|OTTHUMP00000210745
  • Inhibitor of p53 induced apoptosis alpha|Inhibitor of p53-induced apoptosis-alpha|IPIA ALPHA|IPIA-alpha|NDK 6|NDK6_HUMAN|NDP kinase 6|NM23 H6|nm23-H6|NM23H6|Nme6|Nucleoside diphosphate kinase 6|OTTHUMP00000164810|OTTHUMP00000210736|OTTHUMP00000210737|OTTHUMP00000210738|OTTHUMP00000210741|OTTHUMP00000210742|OTTHUMP00000210744|OTTHUMP00000210745
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O75414
  • O75414
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Nucleoside diphosphate kinase 6 protein (1-186AA)
  • Nucleoside diphosphate kinase 6 protein (1-186AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together