Order Entry
Canada
ContactUsLinkComponent
Anti-GNAL Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-GNAL Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-GNAL Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77856-172
  • 77738-535
  • Antigen symbol:
  • GNAL
  • GNAL
  • Antigen synonyms:
  • Adenylate cyclase stimulating G alpha protein|olfactory type|Adenylate cyclase-stimulating G alpha protein|DYT25|G alpha olf|GNAL_HUMAN|Guanine nucleotide binding protein (G protein)|alpha activating activity polypeptide|alpha stimulating activity polypeptide|Guanine nucleotide binding protein G(olf) subunit alpha|Guanine nucleotide-binding protein G(olf) subunit alpha
  • Adenylate cyclase stimulating G alpha protein|olfactory type|Adenylate cyclase-stimulating G alpha protein|DYT25|G alpha olf|GNAL_HUMAN|Guanine nucleotide binding protein (G protein)|alpha activating activity polypeptide|alpha stimulating activity polypeptide|Guanine nucleotide binding protein G(olf) subunit alpha|Guanine nucleotide-binding protein G(olf) subunit alpha
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P38405
  • P38405
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Guanine nucleotide-binding protein G(olf) subunit alpha protein (4-116AA)
  • Recombinant Human Guanine nucleotide-binding protein G(olf) subunit alpha protein (4-116AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together