Anti-HOXA4 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HOXA4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-564
- 77740-283
- Antigen symbol:
- HOXA4
- HOXA4
- Antigen synonyms:
- Dfd like protein|Homeo box A4|Homeobox A4|Homeobox protein Hox-1.4|Homeobox protein Hox-1D|Homeobox protein Hox-A4|Hox 1.4 like protein|hox-1.4|hox-1d|HOX1|HOX1D|HXA4_HUMAN
- Dfd like protein|Homeo box A4|Homeobox A4|Homeobox protein Hox-1.4|Homeobox protein Hox-1D|Homeobox protein Hox-A4|Hox 1.4 like protein|hox-1.4|hox-1d|HOX1|HOX1D|HXA4_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q00056
- Q00056
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Homeobox protein Hox-A4 protein (23-173AA)
- Recombinant Human Homeobox protein Hox-A4 protein (23-173AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together