Anti-IFNGR1 Rabbit Polyclonal Antibody
Catalog # 77738-361
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:IFNGR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat,Monkey
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P15260
- Antigen synonyms:Antiviral Protein Type II|Antiviral protein, type 2|AVP type II|AVP, type 2|CD 119|CD119|CD119 antigen|CDw119|FLJ45734|IFN gamma R alpha|IFN gamma R|IFN gamma R1|IFN-gamma receptor 1|IFN-gamma-R1|IFNG R1|IFNGR 1|IFNGR|Immune interferon receptor 1|Immune interferon receptor for|INGR1_HUMAN|Interferon gamma receptor 1|Interferon gamma receptor alpha chain|Interferon gamma receptor alpha chain precursor
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Antiviral Protein Type II
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA: 1:40000
Type:
Antigen: IFNGR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey