Anti-FAAH Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FAAH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-081
- 77736-390
- Antigen symbol:
- FAAH
- FAAH
- Antigen synonyms:
- Anandamide amidohydrolase 1|Anandamide amidohydrolase|FA2H|FAAH-1|FAAH1_HUMAN|Fatty acid 2-hydroxylase|Fatty acid alpha-hydroxylase|Fatty acid amide hydrolase|Fatty-acid amide hydrolase 1|MGC102823|MGC138146|Oleamide hydrolase 1|Oleamide hydrolase
- Anandamide amidohydrolase 1|Anandamide amidohydrolase|FA2H|FAAH-1|FAAH1_HUMAN|Fatty acid 2-hydroxylase|Fatty acid alpha-hydroxylase|Fatty acid amide hydrolase|Fatty-acid amide hydrolase 1|MGC102823|MGC138146|Oleamide hydrolase 1|Oleamide hydrolase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00519
- O00519
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Fatty-acid amide hydrolase 1 protein (205-342AA)
- Recombinant human Fatty-acid amide hydrolase 1 protein (205-342AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together