Anti-JAM2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: JAM2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77741-392
- 77858-369
- Antigen symbol:
- JAM2
- JAM2
- Antigen synonyms:
- C21orf43|CD 322|CD322|CD322 antigen|JAM 2|JAM B|JAM IT/VE JAM|JAM|vascular endothelial|JAM-2|JAM-B|JAM2_HUMAN|JAMB|Jcam2|Junction adhesion molecule 2|Junction adhesion molecule B|Junctional adhesion molecule 2|Junctional adhesion molecule B|PRO 245|PRO245|Vascular endothelial junction associated molecule|Vascular endothelial junction-associated molecule|VE JAM|VE-JAM|VEJAM
- C21orf43|CD 322|CD322|CD322 antigen|JAM 2|JAM B|JAM IT/VE JAM|JAM|vascular endothelial|JAM-2|JAM-B|JAM2_HUMAN|JAMB|Jcam2|Junction adhesion molecule 2|Junction adhesion molecule B|Junctional adhesion molecule 2|Junctional adhesion molecule B|PRO 245|PRO245|Vascular endothelial junction associated molecule|Vascular endothelial junction-associated molecule|VE JAM|VE-JAM|VEJAM
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P57087
- P57087
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Junctional adhesion molecule B protein (29-238AA)
- Recombinant Human Junctional adhesion molecule B protein (29-238AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together