Anti-LMOD1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: LMOD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-052
- 77742-260
- Antigen symbol:
- LMOD1
- LMOD1
- Antigen synonyms:
- 1D|64 kDa autoantigen 1D|64 kDa autoantigen 1D3|64 kDa autoantigen D1|9530015K06Rik|D1|Leiomodin|Leiomodin-1|LMOD1_HUMAN|muscle form|SM LMOD|SM-Lmod|SMLMOD|Smooth muscle leiomodin|Thyroid associated ophthalmopathy autoantigen|Thyroid-associated ophthalmopathy autoantigen
- 1D|64 kDa autoantigen 1D|64 kDa autoantigen 1D3|64 kDa autoantigen D1|9530015K06Rik|D1|Leiomodin|Leiomodin-1|LMOD1_HUMAN|muscle form|SM LMOD|SM-Lmod|SMLMOD|Smooth muscle leiomodin|Thyroid associated ophthalmopathy autoantigen|Thyroid-associated ophthalmopathy autoantigen
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P29536
- P29536
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Leiomodin-1 protein (29-187AA)
- Recombinant Human Leiomodin-1 protein (29-187AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together