Anti-OGDH Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: OGDH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77745-385
- 77745-386
- Antigen symbol:
- OGDH
- OGDH
- Antigen synonyms:
- 2 oxoglutarate dehydrogenase|2 oxoglutarate dehydrogenase complex component E1|2 oxoglutarate dehydrogenase mitochondrial|2-oxoglutarate dehydrogenase|2-oxoglutarate dehydrogenase complex component E1|AKGDH|Alpha ketoglutarate dehydrogenase|Alpha-ketoglutarate dehydrogenase|E1k|mitochondrial|ODO1_HUMAN|OGDC|OGDC E1|OGDC-E1|OGDH|Oxoglutarate (alpha ketoglutarate) dehydrogenase (lipoamide)|Oxoglutarate decarboxylase|Oxoglutarate dehydrogenase (lipoamide)|Oxoglutarate dehydrogenase (succinyl transferring)
- 2 oxoglutarate dehydrogenase|2 oxoglutarate dehydrogenase complex component E1|2 oxoglutarate dehydrogenase mitochondrial|2-oxoglutarate dehydrogenase|2-oxoglutarate dehydrogenase complex component E1|AKGDH|Alpha ketoglutarate dehydrogenase|Alpha-ketoglutarate dehydrogenase|E1k|mitochondrial|ODO1_HUMAN|OGDC|OGDC E1|OGDC-E1|OGDH|Oxoglutarate (alpha ketoglutarate) dehydrogenase (lipoamide)|Oxoglutarate decarboxylase|Oxoglutarate dehydrogenase (lipoamide)|Oxoglutarate dehydrogenase (succinyl transferring)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q02218
- Q02218
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- 2-oxoglutarate dehydrogenase, mitochondrial protein (41-423AA)
- 2-oxoglutarate dehydrogenase, mitochondrial protein (41-423AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together