Anti-FOXP1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FOXP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-586
- 77737-473
- Antigen symbol:
- FOXP1
- FOXP1
- Antigen synonyms:
- 12CC4|FLJ23741|Fork head related protein like B|Forkhead box P1|Forkhead box protein P1|FOX P1|FOXP 1|FOXP1_HUMAN|Glutamine rich factor 1|hFKH1B|HSPC215|MGC12942|MGC88572|MGC99551|QRF 1|QRF1
- 12CC4|FLJ23741|Fork head related protein like B|Forkhead box P1|Forkhead box protein P1|FOX P1|FOXP 1|FOXP1_HUMAN|Glutamine rich factor 1|hFKH1B|HSPC215|MGC12942|MGC88572|MGC99551|QRF 1|QRF1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H334
- Q9H334
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Forkhead box protein P1 protein (1-114AA)
- Recombinant human Forkhead box protein P1 protein (1-114AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together