Anti-FFAR3 Rabbit Recombinant polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: FFAR3
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-312
- 77737-076
- Antigen symbol:
- FFAR3
- FFAR3
- Antigen synonyms:
- FFA3R|FFAR3_HUMAN|Free fatty acid receptor 3|G protein coupled receptor 41|G-protein coupled receptor 41|gpcr41|GPR41|gpr42
- FFA3R| FFAR3_HUMAN| Free fatty acid receptor 3| G protein coupled receptor 41| G-protein coupled receptor 41| gpcr41| GPR41| gpr42
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14843
- O14843
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Free fatty acid receptor 3 protein (280-346AA)
- Recombinant Human Free fatty acid receptor 3 protein (280-346AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together