Anti-ANO9 Rabbit Polyclonal Antibody
Catalog # 77738-185
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ANO9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:A1A5B4
- Antigen synonyms:ANO9_HUMAN|Anoctamin-9|p53-induced gene 5 protein|PIG5|TMEM16J|TP53I5|Transmembrane protein 16J|Tumor protein p53-inducible protein 5
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Tumor protein p53-inducible protein 5
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA: 1:5000
Type:
Antigen: ANO9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human