Order Entry
Canada
ContactUsLinkComponent
Anti-FUT10 Rabbit Polyclonal Antibody
Anti-FUT10 Rabbit Polyclonal Antibody
  77737-725
New Product
Anti-FUT10 Rabbit Polyclonal Antibody
  77737-725
 :  A22541-100UL
New Product

 

  • Antigen symbol:
    FUT10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse,
    Rat
  • Western blot:
    Yes
  • Size:
    100 µl
  • Form:
    Liquid
  • Gene ID:
    Q6P4F1
  • Antigen synonyms:
    3)-fucosyltransferase 10|Alpha-(1|Alpha-(1,3)-fucosyltransferase 10|Fuc-TX|Fucosyltransferase 10|Fucosyltransferase X|FucT-X|FUT10_HUMAN|Galactoside 3-L-fucosyltransferase 10
  • Storage buffer:
    PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
  • Storage temperature:
    Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Galactoside 3-L-fucosyltransferase 10
  • Purification:
    Antigen affinity purified
  • Packaging:
    Vial

 

 

Type:
Antigen: FUT10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat