Anti-KCNA6 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KCNA6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-444
- 77858-403
- Antigen symbol:
- KCNA6
- KCNA6
- Antigen synonyms:
- HBK 2|HBK2|Human brain potassium channel 2|KCNA 6|KCNA6_HUMAN|Potassium voltage gated channel shaker related subfamily member 6|Potassium voltage gated channel subfamily A member 6|Potassium voltage-gated channel subfamily A member 6|Voltage gated potassium channel protein Kv1.6|Voltage gated potassium channel subunit Kv1.6|Voltage-gated potassium channel HBK2|Voltage-gated potassium channel subunit Kv1.6
- HBK 2|HBK2|Human brain potassium channel 2|KCNA 6|KCNA6_HUMAN|Potassium voltage gated channel shaker related subfamily member 6|Potassium voltage gated channel subfamily A member 6|Potassium voltage-gated channel subfamily A member 6|Voltage gated potassium channel protein Kv1.6|Voltage gated potassium channel subunit Kv1.6|Voltage-gated potassium channel HBK2|Voltage-gated potassium channel subunit Kv1.6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P17658
- P17658
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Potassium voltage-gated channel subfamily A member 6 protein (1-171AA)
- Recombinant Human Potassium voltage-gated channel subfamily A member 6 protein (1-171AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together