Anti-ATP6V1F Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ATP6V1F
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-367
- 77744-368
- Antigen symbol:
- ATP6V1F
- ATP6V1F
- Antigen synonyms:
- Adenosinetriphosphatase 14k chain|ATP6S14|ATPase, H+ transporting, lysosomal 14kDa, V1 subunit F|ATPase, vacuolar, 14 kD|H(+)-transporting two-sector ATPase, 14kD subunit|MGC117321|MGC126037|MGC126038|V-ATPase 14 kDa subunit|V-ATPase subunit F|V-type proton ATPase subunit F|Vacuolar ATP synthase subunit F|Vacuolar proton pump subunit F|VATF|VATF_HUMAN|Vma7
- Adenosinetriphosphatase 14k chain|ATP6S14|ATPase, H+ transporting, lysosomal 14kDa, V1 subunit F|ATPase, vacuolar, 14 kD|H(+)-transporting two-sector ATPase, 14kD subunit|MGC117321|MGC126037|MGC126038|V-ATPase 14 kDa subunit|V-ATPase subunit F|V-type proton ATPase subunit F|Vacuolar ATP synthase subunit F|Vacuolar proton pump subunit F|VATF|VATF_HUMAN|Vma7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16864
- Q16864
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- V-type proton ATPase subunit F protein (1-119AA)
- V-type proton ATPase subunit F protein (1-119AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together