Anti-LGALS4 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LGALS4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-114
- 77858-932
- Antigen symbol:
- LGALS4
- LGALS4
- Antigen synonyms:
- Antigen NY CO 27|Antigen NY-CO-27|Antigen NYCO27|GAL 4|Gal-4|GAL4|Galectin 4|Galectin-4|Galectin4|Homo sapiens galectin4 mRNA complete cds|L 36 lactose binding protein|L-36 lactose-binding protein|L36 lactose binding protein|L36LBP|Lactose binding lectin 4|Lactose-binding lectin 4|Lectin galactoside binding soluble 4|LEG4_HUMAN|LGALS4
- Antigen NY CO 27|Antigen NY-CO-27|Antigen NYCO27|GAL 4|Gal-4|GAL4|Galectin 4|Galectin-4|Galectin4|Homo sapiens galectin4 mRNA complete cds|L 36 lactose binding protein|L-36 lactose-binding protein|L36 lactose binding protein|L36LBP|Lactose binding lectin 4|Lactose-binding lectin 4|Lectin galactoside binding soluble 4|LEG4_HUMAN|LGALS4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P56470
- P56470
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Galectin-4 protein (1-323AA)
- Recombinant Human Galectin-4 protein (1-323AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together