Anti-MAGEA4 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAGEA4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-567
- 77859-314
- Antigen symbol:
- MAGEA4
- MAGEA4
- Antigen synonyms:
- Cancer/testis antigen 1.4|CT1.4|MAGA4_HUMAN|MAGE 4|MAGE 4 antigen|MAGE 41|MAGE 41 antigen|MAGE 4A|MAGE 4B|MAGE X2|MAGE X2 antigen|MAGE-4 antigen|MAGE-41 antigen|MAGE-X2 antigen|MAGE4|MAGE41|MAGE4A|MAGE4B|MAGEA 4|Melanoma antigen family A 4|Melanoma associated antigen 4|Melanoma-associated antigen 4|MGC21336
- Cancer/testis antigen 1.4|CT1.4|MAGA4_HUMAN|MAGE 4|MAGE 4 antigen|MAGE 41|MAGE 41 antigen|MAGE 4A|MAGE 4B|MAGE X2|MAGE X2 antigen|MAGE-4 antigen|MAGE-41 antigen|MAGE-X2 antigen|MAGE4|MAGE41|MAGE4A|MAGE4B|MAGEA 4|Melanoma antigen family A 4|Melanoma associated antigen 4|Melanoma-associated antigen 4|MGC21336
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P43358
- P43358
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Melanoma-associated antigen 4 protein (1-270AA)
- Recombinant Human Melanoma-associated antigen 4 protein (1-270AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together