Anti-IL2RG Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IL2RG
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-055
- 77741-005
- Antigen symbol:
- IL2RG
- IL2RG
- Antigen synonyms:
- CD132| CD132 antigen| CIDX| common cytokine receptor gamma chain| Cytokine receptor common subunit gamma| Gamma C| gamma(c)| gammaC| IL-2 receptor subunit gamma| IL-2R gamma chain| IL-2R subunit gamma| IL-2RG| Il2rg| IL2RG_HUMAN| IMD4| interleukin 2 receptor| gamma| Interleukin-2 receptor subunit gamma| p64| SCIDX| SCIDX1
- CD132|CD132 antigen|CIDX|common cytokine receptor gamma chain|Cytokine receptor common subunit gamma|Gamma C|gamma(c)|gammaC|IL-2 receptor subunit gamma|IL-2R gamma chain|IL-2R subunit gamma|IL-2RG|Il2rg|IL2RG_HUMAN|IMD4|interleukin 2 receptor|gamma|Interleukin-2 receptor subunit gamma|p64|SCIDX|SCIDX1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31785
- P31785
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Cytokine receptor common subunit gamma protein (75-230AA)
- Recombinant Human Cytokine receptor common subunit gamma protein (75-230AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together