Anti-GCNT3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500 to 1:5000, IHC:1:20 to 1:200
Type:
Antigen: GCNT3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-874
- 77738-089
- Antigen symbol:
- GCNT3
- Antigen synonyms:
- 2010013H22Rik|2210021I22Rik|2210401J11Rik|3-galactosyl-O-glycosyl-glycoprotein beta-1|6-N-acetylglucosaminyltransferase 3|6-N-acetylglucosaminyltransferase|Beta 1 3 galactosyl O glycosyl glycoprotein beta 1 6 N acetylglucosaminyltransferase 3|Beta-1|Beta1 6 N acetylglucosaminyltransferase 3|beta1 6 N acetylglucosaminyltransferase|C2/4GnT|C24GNT|C2GnT M|C2GnT mucin type|C2GnT-M|C2GnT-mucin type|C2GnT2|C2GNTM|Core 2 beta 1 6 N acetylglucosaminyltransferase II|Core 2/core 4 beta 1 6 N acetylglucosaminyltransferase|Core 2/core 4 beta-1|dI/C2/C4GnT|EC 2.4.1.102|EC 2.4.1.150|GCNT3|GCNT3_HUMAN|Glucosaminyl (N acetyl) transferase 3|Glucosaminyl (N acetyl) transferase 3 mucin type|glucosaminyl (N-acetyl) transferase 3|mucin type|GnT M|GNTM|hC2GnT M|hC2GnT-M|Mucus-type core 2 beta-1,6-N-acetylglucosaminyltransferase|OTTHUMP00000163601
- 2010013H22Rik|2210021I22Rik|2210401J11Rik|3-galactosyl-O-glycosyl-glycoprotein beta-1|6-N-acetylglucosaminyltransferase 3|6-N-acetylglucosaminyltransferase|Beta 1 3 galactosyl O glycosyl glycoprotein beta 1 6 N acetylglucosaminyltransferase 3|Beta-1|Beta1 6 N acetylglucosaminyltransferase 3|beta1 6 N acetylglucosaminyltransferase|C2/4GnT|C24GNT|C2GnT M|C2GnT mucin type|C2GnT-M|C2GnT-mucin type|C2GnT2|C2GNTM|Core 2 beta 1 6 N acetylglucosaminyltransferase II|Core 2/core 4 beta 1 6 N acetylglucosaminyltransferase|Core 2/core 4 beta-1|dI/C2/C4GnT|EC 2.4.1.102|EC 2.4.1.150|GCNT3|GCNT3_HUMAN|Glucosaminyl (N acetyl) transferase 3|Glucosaminyl (N acetyl) transferase 3 mucin type|glucosaminyl (N-acetyl) transferase 3|mucin type|GnT M|GNTM|hC2GnT M|hC2GnT-M|Mucus-type core 2 beta-1,6-N-acetylglucosaminyltransferase|OTTHUMP00000163601
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- O95395
- O95395
- Isotype:
- IgG
- Western blot:
- Yes
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 3 protein (250-419AA)
- Recombinant Human Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 3 protein (250-419AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together