Anti-YWHAZ Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: YWHAZ
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-812
- 77743-196
- Antigen symbol:
- YWHAZ
- YWHAZ
- Antigen synonyms:
- 14 3 3 delta|14 3 3 protein zeta/delta|14 3 3 protein/cytosolic phospholipase A2|14 3 3 zeta|14-3-3 protein zeta/delta|1433Z_HUMAN|Epididymis luminal protein 4|Epididymis secretory protein Li 3|HEL S 3|HEL4|KCIP-1|KCIP1|MGC111427|MGC126532|MGC138156|Phospholipase A2|Protein kinase C inhibitor protein 1|Tyrosine 3 monooxygenase/tryptophan 5 monooxygenase activation protein|delta polypeptide|zeta|zeta polypeptide|Tyrosine 3/tryptophan 5 monooxygenase activation protein|YWHAD|YWHAZ
- HEL4|Tyrosine 3/tryptophan 5 monooxygenase activation protein|zeta|YWHAD|Phospholipase A2|MGC126532|delta polypeptide|KCIP1|1433Z_HUMAN|YWHAZ|zeta polypeptide|Epididymis luminal protein 4|Protein kinase C inhibitor protein 1|MGC111427|14 3 3 protein/cytosolic phospholipase A2|14 3 3 delta|14 3 3 zeta|14-3-3 protein zeta/delta|HEL S 3|KCIP-1|Epididymis secretory protein Li 3|14 3 3 protein zeta/delta|MGC138156|Tyrosine 3 monooxygenase/tryptophan 5 monooxygenase activation protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P63104
- P63104
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human 14-3-3 protein zeta/delta protein (133-212AA)
- Recombinant Human 14-3-3 protein zeta/delta protein (133-212AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together