Anti-FKBP3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FKBP3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-239
- 77855-437
- Antigen symbol:
- FKBP3
- FKBP3
- Antigen synonyms:
- 25 kDa FK506-binding protein|25 kDa FKBP|FK506 binding protein 25 T cell|FK506 binding protein 25|T cell|25-KD|FK506 binding protein 3 (25kD)|FK506-binding protein 3|FKBP 25|FKBP 3|FKBP-25|FKBP-3|FKBP25|FKBP3_HUMAN|Immunophilin FKBP25|Peptidyl-prolyl cis-trans isomerase PPIase|PPIase Rapamycin binding protein|Rapamycin-selective 25 kDa immunophilin|Rotamase
- 25 kDa FK506-binding protein|25 kDa FKBP|FK506 binding protein 25 T cell|FK506 binding protein 25|T cell|25-KD|FK506 binding protein 3 (25kD)|FK506-binding protein 3|FKBP 25|FKBP 3|FKBP-25|FKBP-3|FKBP25|FKBP3_HUMAN|Immunophilin FKBP25|Peptidyl-prolyl cis-trans isomerase PPIase|PPIase Rapamycin binding protein|Rapamycin-selective 25 kDa immunophilin|Rotamase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q00688
- Q00688
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Peptidyl-prolyl cis-trans isomerase FKBP3 protein (2-224AA)
- Recombinant Human Peptidyl-prolyl cis-trans isomerase FKBP3 protein (2-224AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together