Anti-MRPS18C Rabbit Recombinant polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MRPS18C
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-992
- 77743-461
- Antigen symbol:
- MRPS18C
- MRPS18C
- Antigen synonyms:
- CGI-134|28S ribosomal protein S18c|mitochondrial|MRP-S18-c|Mrps18-c|S18mt-c|28S ribosomal protein S18-1|MRP-S18-1|Mitochondrial small ribosomal subunit protein bS18c|Mitochondrial small ribosomal subunit protein bS18m
- CGI-134| 28S ribosomal protein S18c| mitochondrial| MRP-S18-c| Mrps18-c| S18mt-c| 28S ribosomal protein S18-1| MRP-S18-1| Mitochondrial small ribosomal subunit protein bS18c| Mitochondrial small ribosomal subunit protein bS18m
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3D5
- Q9Y3D5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human 28S ribosomal protein S18c, mitochondrial protein (1-142AA)
- Recombinant Human 28S ribosomal protein S18c, mitochondrial protein (1-142AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together