Anti-GPKOW Rabbit Polyclonal Antibody
Catalog # 77738-756
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:GPKOW
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q92917
- Antigen synonyms:G patch domain and KOW motifs|G patch domain and KOW motifs containing protein|G patch domain and KOW motifs-containing protein|G patch domain containing 5|G patch domain containing protein 5|G patch domain-containing protein 5|GPATC5|GPATCH5|GPKOW_HUMAN|Protein MOS2 homolog|Protein T54|SPP2|T54|T54 protein
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:T54 protein
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: GPKOW
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat