Anti-H2AFY Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: H2AFY
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-685
- 77739-366
- Antigen symbol:
- H2AFY
- Antigen synonyms:
- Core histone macro h2a.1|Core histone macro-H2A.1|H2A histone family member Y|H2A.y|H2A/y|H2AF12M|H2AFJ|H2afy|H2AY_HUMAN|Histone H2A.Y|Histone macroH2A1|Histone macroH2A1.1|Histone macroH2A1.2|Macroh2a1|MACROH2A1.1|MacroH2A1.2|Medulloblastoma antigen MU MB 50.205|Medulloblastoma antigen MU-MB-50.205|mH2a|mH2A1
- Core histone macro h2a.1|Core histone macro-H2A.1|H2A histone family member Y|H2A.y|H2A/y|H2AF12M|H2AFJ|H2afy|H2AY_HUMAN|Histone H2A.Y|Histone macroH2A1|Histone macroH2A1.1|Histone macroH2A1.2|Macroh2a1|MACROH2A1.1|MacroH2A1.2|Medulloblastoma antigen MU MB 50.205|Medulloblastoma antigen MU-MB-50.205|mH2a|mH2A1
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- O75367
- O75367
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human Core histone macro-H2A.1 protein (123-372AA)
- Recombinant Human Core histone macro-H2A.1 protein (123-372AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together