Anti-LRRN2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: LRRN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77859-165
- 77742-387
- Antigen symbol:
- LRRN2
- LRRN2
- Antigen synonyms:
- LRRN2| GAC1| LRRN5| UNQ256/PRO293Leucine-rich repeat neuronal protein 2| Glioma amplified on chromosome 1 protein| Leucine-rich repeat neuronal protein 5
- LRRN2|GAC1|LRRN5|UNQ256/PRO293Leucine-rich repeat neuronal protein 2|Glioma amplified on chromosome 1 protein|Leucine-rich repeat neuronal protein 5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75325
- O75325
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Leucine-rich repeat neuronal protein 2 protein (201-500AA)
- Recombinant Human Leucine-rich repeat neuronal protein 2 protein (201-500AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together