Order Entry
Canada
ContactUsLinkComponent
Anti-KRT6A Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur:  AFG Bioscience
New Product
Certificats
undefined
Anti-KRT6A Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur:  AFG Bioscience
New Product

Spécifications

  • Catalog No:
  • 77741-889
  • 77858-752
  • Antigen symbol:
  • KRT6A
  • KRT6A
  • Antigen synonyms:
  • CK-6A|CK-6D|CK6A|CK6C|CK6D|Cytokeratin-6A|Cytokeratin-6D|K2C6A_HUMAN|K6A|K6C|K6D|keratin 6A|Keratin|type II cytoskeletal 6A|Keratin-6A|KRT6C|KRT6D|Type-II keratin Kb6
  • CK-6A|CK-6D|CK6A|CK6C|CK6D|Cytokeratin-6A|Cytokeratin-6D|K2C6A_HUMAN|K6A|K6C|K6D|keratin 6A|Keratin|type II cytoskeletal 6A|Keratin-6A|KRT6C|KRT6D|Type-II keratin Kb6
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P02538
  • P02538
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Keratin, type II cytoskeletal 6A protein (2-564AA)
  • Recombinant Human Keratin, type II cytoskeletal 6A protein (2-564AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Spécifications

Frequently Bought Together