Anti-LMOD1 Rabbit Polyclonal Antibody
77742-261
New Product
- Antigen symbol:LMOD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P29536
- Antigen synonyms:1D|64 kDa autoantigen 1D|64 kDa autoantigen 1D3|64 kDa autoantigen D1|9530015K06Rik|D1|Leiomodin|Leiomodin-1|Lmod1|LMOD1_HUMAN|muscle form|SM LMOD|SM-Lmod|SMLMOD|Smooth muscle leiomodin|Thyroid associated ophthalmopathy autoantigen|Thyroid-associated ophthalmopathy autoantigen
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Thyroid-associated ophthalmopathy autoantigen
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: LMOD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat