Anti-IFNA10 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: IFNA10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77740-665
- 77857-869
- Antigen symbol:
- IFNA10
- IFNA10
- Antigen synonyms:
- IFN-alpha-10|IFN10_HUMAN|IFNA10|Interferon alpha 6L|Interferon alpha C|Interferon alpha-10|Interferon alpha-6L|Interferon alpha-C|interferon, alpha 10|LeIF C|MGC119878|MGC119879
- IFN-alpha-10|IFN10_HUMAN|IFNA10|Interferon alpha 6L|Interferon alpha C|Interferon alpha-10|Interferon alpha-6L|Interferon alpha-C|interferon, alpha 10|LeIF C|MGC119878|MGC119879
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P01566
- P01566
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant huamn Interferon alpha-10 protein (24-189AA)
- Recombinant huamn Interferon alpha-10 protein (24-189AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together