Anti-EPHA4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: EPHA4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77854-793
- 77735-991
- Antigen symbol:
- EPHA4
- EPHA4
- Antigen synonyms:
- Cek 8|CEK8|EK8|eph receptor a4|EPH-like kinase 8|EPHA4_HUMAN|Ephrin type-A receptor 4|HEK 8|hEK8|Receptor protein-tyrosine kinase HEK8|Sek 1|SEK|TYRO 1 protein tyrosine kinase|TYRO1|Tyrosine-protein kinase receptor SEK|Tyrosine-protein kinase TYRO1
- Cek 8|CEK8|EK8|eph receptor a4|EPH-like kinase 8|EPHA4_HUMAN|Ephrin type-A receptor 4|HEK 8|hEK8|Receptor protein-tyrosine kinase HEK8|Sek 1|SEK|TYRO 1 protein tyrosine kinase|TYRO1|Tyrosine-protein kinase receptor SEK|Tyrosine-protein kinase TYRO1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P54764
- P54764
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ephrin type-A receptor 4 protein (345-545AA)
- Recombinant Human Ephrin type-A receptor 4 protein (345-545AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together