Anti-SLC39A2 Rabbit Polyclonal Antibody
Catalog # 77737-723
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:SLC39A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NP94
- Antigen synonyms:hZIP2|6A1|Eti-1|Eti1|MGC119190|OTTHUMP00000164402|S39A2_HUMAN|SLC39A2|Solute carrier family 39 (zinc transporter) member 2|Solute carrier family 39 member 2|Zinc transporter ZIP2|ZIP 2|ZIP-2|ZIP2|Zrt and Irt like protein 2|Zrt- and Irt-like protein 2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:hZIP2
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:10000
Type:
Antigen: SLC39A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human