Order Entry
Canada
ContactUsLinkComponent
Anti-HSPA9 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-HSPA9 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-HSPA9 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77740-484
  • 77857-725
  • Antigen symbol:
  • HSPA9
  • HSPA9
  • Antigen synonyms:
  • 75 kDa glucose regulated protein|75 kDa glucose-regulated protein|CSA|Glucose Regulated Protein|Grp 75|GRP-75|GRP75|GRP75_HUMAN|Heat shock 70 kDa protein 9|Heat shock 70kD protein 9|heat shock 70kDa protein 9|Heat shock 70kDa protein 9B|Heat shock protein 74 kDa A|Heat shock protein A|Heat shock protein cognate 74|Hsc74|Hsp74|Hsp74a|HSPA9|Hspa9a|HSPA9B|MGC4500|mitochondrial|Mortalin 2|Mortalin|Mortalin perinuclear|Mortalin2|MOT 2|MOT|MOT2|Mthsp70|p66 mortalin|P66 MOT|PBP74|Peptide binding protein 74|Peptide-binding protein 74|Stress 70 protein mitochondrial|Stress 70 protein mitochondrial precursor|Stress-70 protein
  • 75 kDa glucose regulated protein| 75 kDa glucose-regulated protein| CSA| Glucose Regulated Protein| Grp 75| GRP-75| GRP75| GRP75_HUMAN| Heat shock 70 kDa protein 9| Heat shock 70kD protein 9| heat shock 70kDa protein 9| Heat shock 70kDa protein 9B| Heat shock protein 74 kDa A| Heat shock protein A| Heat shock protein cognate 74| Hsc74| Hsp74| Hsp74a| HSPA9| Hspa9a| HSPA9B| MGC4500| mitochondrial| Mortalin 2| Mortalin| Mortalin perinuclear| Mortalin2| MOT 2| MOT| MOT2| Mthsp70| p66 mortalin| P66 MOT| PBP74| Peptide binding protein 74| Peptide-binding protein 74| Stress 70 protein mitochondrial| Stress 70 protein mitochondrial precursor| Stress-70 protein
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P38646
  • P38646
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Stress-70 protein, mitochondrial protein (458-679AA)
  • Recombinant Human Stress-70 protein, mitochondrial protein (458-679AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together