Anti-RAPGEF1 Rabbit Polyclonal Antibody
Catalog # 77737-592
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:RAPGEF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q13905
- Antigen synonyms:C3G|C3G protein|CRK SH3 binding GNRP|CRK SH3-binding GNRP|DKFZp781P1719|GRF 2|GRF2|Guanine nucleotide releasing factor 2 (specific for crk proto oncogene)|Guanine nucleotide releasing factor 2|Guanine nucleotide-releasing factor 2|OTTHUMP00000064558|Protein C3G|Rap guanine nucleotide exchange factor (GEF) 1|Rap guanine nucleotide exchange factor 1|RAPGEF 1|RPGF1_HUMAN
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:C3G
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:100 to 1:300, ELISA:1:5000
Type:
Antigen: RAPGEF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human