Anti-LTF Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LTF
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-454
- 77737-263
- Antigen symbol:
- LTF
- LTF
- Antigen synonyms:
- GIG12|Growth inhibiting protein 12|HLF2|Kaliocin 1|Lactoferrin|Lactoferroxin-C|Lactotransferrin|LF|LTF|Neutrophil lactoferrin|Talalactoferrin|TRFL_HUMAN
- GIG12|Growth inhibiting protein 12|HLF2|Kaliocin 1|Lactoferrin|Lactoferroxin-C|Lactotransferrin|LF|LTF|Neutrophil lactoferrin|Talalactoferrin|TRFL_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P02788
- P02788
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Human Lactotransferrin protein (Native Protein)
- Human Lactotransferrin protein (Native Protein)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together