Anti-LDB3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: LDB3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-876
- 77742-039
- Antigen symbol:
- LDB3
- LDB3
- Antigen synonyms:
- CMD1C|CYPHER|HGNC:15710|KIAA01613|KIAA0613|LDB3_HUMAN|LDB3Z1|LDB3Z4|LIM domain binding 3|LIM domain binding protein 3|LIM domain-binding protein 3|LVNC3|ORACLE|PDLIM6|PDZ and LIM domain 6|Protein cypher|Z band alternatively spliced PDZ motif|Z band alternatively spliced PDZ motif protein|Z-band alternatively spliced PDZ-motif protein|ZASP
- CMD1C|CYPHER|HGNC:15710|KIAA01613|KIAA0613|LDB3_HUMAN|LDB3Z1|LDB3Z4|LIM domain binding 3|LIM domain binding protein 3|LIM domain-binding protein 3|LVNC3|ORACLE|PDLIM6|PDZ and LIM domain 6|Protein cypher|Z band alternatively spliced PDZ motif|Z band alternatively spliced PDZ motif protein|Z-band alternatively spliced PDZ-motif protein|ZASP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75112
- O75112
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human LIM domain-binding protein 3 protein (1-283AA)
- Recombinant human LIM domain-binding protein 3 protein (1-283AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together