Anti-HIST1H2AG Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:10 to 1:100
Type:
Antigen: HIST1H2AG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77739-790
- 77857-005
- Antigen symbol:
- HIST1H2AG
- HIST1H2AG
- Antigen synonyms:
- H2AC11|H2AFP|HIST1H2AG||H2AC13|H2AFC|HIST1H2AI|H2AC15|H2AFD|HIST1H2AK|H2AC16|H2AFI|HIST1H2AL|H2AC17|H2AFN|HIST1H2AMHistone H2A type 1|H2A.1|Histone H2A/ptl
- H2AC11|H2AFP|HIST1H2AG||H2AC13|H2AFC|HIST1H2AI|H2AC15|H2AFD|HIST1H2AK|H2AC16|H2AFI|HIST1H2AL|H2AC17|H2AFN|HIST1H2AMHistone H2A type 1|H2A.1|Histone H2A/ptl
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P0C0S8
- P0C0S8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Peptide sequence around site of Lys (118) derived from Human Histone H2A type 1
- Peptide sequence around site of Lys (118) derived from Human Histone H2A type 1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together