Anti-NDUFS2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: NDUFS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse
Specifications
- Catalog No:
- 77744-212
- 77744-213
- Antigen symbol:
- NDUFS2
- NDUFS2
- Antigen synonyms:
- CI 49|CI 49kD|CI-49kD|Complex 1, mitochondrial respiratory chain, 49 KD subunit|Complex I 49kD|Complex I 49kDa subunit|Complex I-49kD|mitochondrial|NADH dehydrogenase (ubiquinone) Fe S protein 2 49kDa|NADH dehydrogenase (ubiquinone) Fe S protein 2, 49kDa (NADH coenzyme Q reductase)|NADH dehydrogenase [ubiquinone] iron sulfur protein 2, mitochondrial|NADH dehydrogenase [ubiquinone] iron-sulfur protein 2|NADH ubiquinone oxidoreductase 49 kDa subunit|NADH ubiquinone oxidoreductase NDUFS2 subunit|NADH-ubiquinone oxidoreductase 49 kDa subunit|NADH:ubiquinone oxidoreductase core subunit S2|NDUS2_HUMAN
- CI 49|CI 49kD|CI-49kD|Complex 1, mitochondrial respiratory chain, 49 KD subunit|Complex I 49kD|Complex I 49kDa subunit|Complex I-49kD|mitochondrial|NADH dehydrogenase (ubiquinone) Fe S protein 2 49kDa|NADH dehydrogenase (ubiquinone) Fe S protein 2, 49kDa (NADH coenzyme Q reductase)|NADH dehydrogenase [ubiquinone] iron sulfur protein 2, mitochondrial|NADH dehydrogenase [ubiquinone] iron-sulfur protein 2|NADH ubiquinone oxidoreductase 49 kDa subunit|NADH ubiquinone oxidoreductase NDUFS2 subunit|NADH-ubiquinone oxidoreductase 49 kDa subunit|NADH:ubiquinone oxidoreductase core subunit S2|NDUS2_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75306
- O75306
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial protein (297-427AA)
- NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial protein (297-427AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together