Anti-IL4 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: IL4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77858-070
- 77741-023
- Antigen symbol:
- IL4
- IL4
- Antigen synonyms:
- Interleukin 4 variant 2| B-cell stimulatory factor 1| B Cell Stimulatory Factor 1| Lymphocyte stimulatory factor 1| BSF1 | B cellIgG differentiation factor| BCGF 1| Interleukin 4| BCGF1| IL-4| Pitrakinra| Il4e12| IL 4| isoform 1 | MGC79402| IL4_HUMAN| Interleukin-4| BSF-1|IgG1 induction factor|B cell growth factor 1| IL4| Binetrakin
- B cell growth factor 1|B cell IgG differentiation factor|B Cell Stimulatory Factor 1|B-cell stimulatory factor 1|BCGF 1|BCGF1|Binetrakin|BSF-1|BSF1 |IGG1 induction factor|IL 4|IL-4|IL4|IL4_HUMAN|Il4e12|Interleukin 4|Interleukin 4 variant 2|isoform 1 |Interleukin-4|Lymphocyte stimulatory factor 1|MGC79402|Pitrakinra
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P05112
- P05112
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Interleukin-4 protein (25-153AA)
- Recombinant Human Interleukin-4 protein (25-153AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together