Anti-MRPS18A Rabbit Polyclonal Antibody
Catalog # 77743-456
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MRPS18A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NVS2
- Antigen synonyms:39S ribosomal protein S18a| mitochondrial| MRP-S18-a| S18mt-a| 39S ribosomal protein S18-3| MRP-S18-3| Mitochondrial large ribosomal subunit protein bS18a| Mitochondrial large ribosomal subunit protein mL66
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:Shipped on Blue ice
- Immunogen:Mitochondrial large ribosomal subunit protein mL66
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MRPS18A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat