Anti-KPNA4 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KPNA4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-700
- 77741-822
- Antigen symbol:
- KPNA4
- KPNA4
- Antigen synonyms:
- FLJ31113| IMA3_HUMAN| Importin alpha 3| Importin alpha 4 subunit| Importin alpha Q1| Importin subunit alpha 3| Importin subunit alpha-3| IPO A3| IPOA 3| IPOA3| Karyopherin alpha 4 (importin alpha 3)| Karyopherin alpha 4| Karyopherin alpha 4 importin alpha 3| Karyopherin alpha 4 subunit| Karyopherin subunit alpha 4| Karyopherin subunit alpha-4| KPNA 4| Kpna4| MGC12217| MGC26703| OTTHUMP00000213880| QIP 1| Qip1| Qip1 protein| SRP 3| SRP3
- FLJ31113|IMA3_HUMAN|Importin alpha 3|Importin alpha 4 subunit|Importin alpha Q1|Importin subunit alpha 3|Importin subunit alpha-3|IPO A3|IPOA 3|IPOA3|Karyopherin alpha 4 (importin alpha 3)|Karyopherin alpha 4|Karyopherin alpha 4 importin alpha 3|Karyopherin alpha 4 subunit|Karyopherin subunit alpha 4|Karyopherin subunit alpha-4|KPNA 4|Kpna4|MGC12217|MGC26703|OTTHUMP00000213880|QIP 1|Qip1|Qip1 protein|SRP 3|SRP3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00629
- O00629
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Importin subunit alpha-3 protein (51-239AA)
- Recombinant Human Importin subunit alpha-3 protein (51-239AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together