Anti-FGF4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FGF4
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-136
- 77855-367
- Antigen symbol:
- FGF4
- Antigen synonyms:
- FGF-4|Fgf4|FGF4_HUMAN|Fibroblast growth factor 4|fibroblast growth factor 4 splice isoform|HBGF-4|HBGF4|Heparin secretory-transforming protein 1|Heparin-binding growth factor 4|Hst|HST-1|HST1|HSTF-1|HSTF1|Human stomach cancer transforming factor from FGF related oncogene|K FGF|Kaposi Sarcoma Oncogene|KFGF|KS3|Oncogene HST|Transforming protein KS3
- FGF-4|Fgf4|FGF4_HUMAN|Fibroblast growth factor 4|fibroblast growth factor 4 splice isoform|HBGF-4|HBGF4|Heparin secretory-transforming protein 1|Heparin-binding growth factor 4|Hst|HST-1|HST1|HSTF-1|HSTF1|Human stomach cancer transforming factor from FGF related oncogene|K FGF|Kaposi Sarcoma Oncogene|KFGF|KS3|Oncogene HST|Transforming protein KS3
- Clonality:
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P08620
- P08620
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Peptide sequence from Human Fibroblast growth factor 4 protein (182-200AA)
- Peptide sequence from Human Fibroblast growth factor 4 protein (182-200AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together