Order Entry
Canada
ContactUsLinkComponent
Anti-FOXN1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-FOXN1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-FOXN1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77855-568
  • 77737-448
  • Antigen symbol:
  • FOXN1
  • FOXN1
  • Antigen synonyms:
  • FKHL20|Forkhead box N1|Forkhead box protein N1|FOXN 1|FOXN1_HUMAN|RONU|Rowett nude|Transcription factor winged-helix nude|WHN|Winged helix nude|Winged-helix transcription factor nude
  • FKHL20|Forkhead box N1|Forkhead box protein N1|FOXN 1|FOXN1_HUMAN|RONU|Rowett nude|Transcription factor winged-helix nude|WHN|Winged helix nude|Winged-helix transcription factor nude
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O15353
  • O15353
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Forkhead box protein N1 protein (1-160AA)
  • Recombinant Human Forkhead box protein N1 protein (1-160AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, protein G purified
  • >95%, protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together